missing translation for 'onlineSavingsMsg'
Learn More

Pardoprunox hydrochloride, MedChemExpress

Product Code. 16426616
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16426616

Brand: Medchem Express HY14958A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Pardoprunox (SLV-308) hydrochloride is a partial dopamine D2 and D3 receptor partial agonist and a serotonin 5-HT1A receptor agonist, with pEC50s of 8, 9.2, and 6.3, respectively.

Pardoprunox (SLV-308) hydrochloride is a partial dopamine D2 and D3 receptor partial agonist and a serotonin 5-HT1A receptor agonist, with pEC50s of 8, 9.2, and 6.3, respectively.

Specifications

Chemical Name or Material Pardoprunox hydrochloride
CAS 269718-83-4
Color Earth Yellow
Purity Grade Notes Research
Molecular Formula C12H16ClN3O2
Quantity 100 mg
Synonym SLV-308 hydrochloride DU-126891 hydrochloride
Solubility Information DMSO : 150 mg/mL (556.11 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES O=C1OC2=C(N3CCN(C)CC3)C=CC=C2N1.[H]Cl
Molecular Weight (g/mol) 269.73
Formula Weight 269.73
Percent Purity 98.24%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.