missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Anti-Human CD235a (HIR2), Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 65174-1-IG-100UG
This item is not returnable.
View return policy
Description
The CD235a antibody from Proteintech is a mouse monoclonal antibody generated with synthetic peptide (human, n-terminal) of human CD235a. This antibody recognizes human antigen. The CD235a antibody has been validated for the following applications: FC analysis.Specifications
| CD235a | |
| Monoclonal | |
| 0.5 mg/mL | |
| CD235a; GPA; GPErik; GPSAT; GpMiIII; HGpMiIII; HGpMiV; HGpMiX; HGpMiXI; HGpSta(C); MN; MNS | |
| Synthetic peptide (Human, N-terminal) | |
| 100 μg | |
| Primary | |
| Human | |
| Flow Antibody | |
| IgG2b κ |
| Flow Cytometry | |
| HIR2 | |
| Unconjugated | |
| Mouse | |
| Affinity purification | |
| RUO | |
| 2993 | |
| Store at 2-8°C. Stable for one year after shipment. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction