missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytokeratin 15 Mouse anti-Human, Clone: KRT15/2959, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-07620-100ug
This item is not returnable.
View return policy
Description
Cytokeratin 15 Monoclonal specifically detects Cytokeratin 15 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Protein Array.
Specifications
| Cytokeratin 15 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Western Blot 1 to 2 μg/mL, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1 to 2 μg/million cells, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen 1 to 2 μg/mL, Protein Array | |
| CK15, CK-15, cytokeratin 15, cytokeratin-15, K15keratin-15, basic, K1CO, keratin 15, keratin, type I cytoskeletal 15, Keratin-15, keratin-15, beta, KRTB, type I cytoskeletal 15 | |
| Recombinant full-length human Cytokeratin 15 protein | |
| 100 μg | |
| Cell Biology, Cellular Markers | |
| 3866 | |
| Store at 4°C. | |
| IgG2c κ |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| KRT15/2959 | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction