missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human B7-H6 Alexa Fluor 488-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems FAB7144G-100UG
This item is not returnable.
View return policy
Description
B7-H6 Monoclonal specifically detects B7-H6 in Human samples. It is validated for Flow Cytometry.
Specifications
| B7-H6 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| Q68D85 | |
| NCR3LG1 | |
| Mouse myeloma cell line NS0-derived recombinant human B7-H6 Met1-Ser262 Accession # Q68D85 | |
| RUO | |
| 374383 | |
| Human | |
| IgG1 |
| Flow Cytometry | |
| 875001 | |
| Alexa Fluor 488 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| B7 homolog 6, B7H6, B7-H6, DKFZp686I21167, DKFZp686O24166, NCR3LG1 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Detects human B7-H6 in ELISA. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction