missing translation for 'onlineSavingsMsg'
Learn More
Learn More
beta II Tubulin Antibody (OTI1H3), Novus Biologicals Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-44660
This item is not returnable.
View return policy
Description
beta II Tubulin Monoclonal antibody specifically detects beta II Tubulin in Human,Mouse,Rat samples. It is validated for Western Blot
Specifications
| beta II Tubulin | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized powder ( 1X PBS, pH 7.3,) | |
| Full length human recombinant protein of human TUBB2A(NP_001060) produced in HEK293T cell. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG2b |
| Western Blot | |
| OTI1H3 | |
| Western Blot 1:4000 | |
| Mouse | |
| >95% | |
| RUO | |
| For reconstitution, we recommend adding 100uL distilled water to a final antibody concentration of about 1 mg/mL. To use this carrier-free antibody for conjugation experiment, we strongly recommend performing another round of desalting process. | |
| Store at -20 degrees C. Avoid freeze/thaw cycles. | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction