missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NUDT4 Antibody (OTI1C2), Novus Biologicals Biologicals™
Mouse Monoclonal Antibody
Marca: Novus Biologicals NBP3-44808
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
NUDT4 Monoclonal antibody specifically detects NUDT4 in Human,Mouse,Rat samples. It is validated for Western Blot
Especificaciones
| NUDT4 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized powder ( 1X PBS, pH 7.3,) | |
| Mouse | |
| >95% | |
| RUO | |
| Primary | |
| For reconstitution, we recommend adding 100uL distilled water to a final antibody concentration of about 1 mg/mL. To use this carrier-free antibody for conjugation experiment, we strongly recommend performing another round of desalting process. | |
| Store at -20 degrees C. Avoid freeze/thaw cycles. | |
| Lyophilized |
| Western Blot | |
| OTI1C2 | |
| Western Blot 1:2000 | |
| Diadenosine 5'-5'''-P1, diphosphoinositol polyphosphate phosphohydrolase 2, diphosphoinositol polyphosphate phosphohydrolase type 2, DIPP-2, DIPP2alpha, DIPP2beta, DIPP2HDCMB47P, EC 3.6.1.-, EC 3.6.1.52, KIAA0487DKFZp686I1281, Nucleoside diphosphate-linked moiety X motif 4, nudix (nucleoside diphosphate linked moiety X)-type motif 4, Nudix motif 4, P6-hexaphosphate hydrolase 2 | |
| Full length human recombiant protein of human NUDT4 (NP_061967) produced in E.coli. | |
| 100 μg | |
| Signal Transduction | |
| 11163 | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG2a |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido