missing translation for 'onlineSavingsMsg'
Learn More
Learn More
POLR2F Antibody (OTI2F2), Novus Biologicals Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-44696
This item is not returnable.
View return policy
Description
POLR2F Monoclonal antibody specifically detects POLR2F in Human,Mouse,Rat samples. It is validated for Western Blot
Specifications
| POLR2F | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized powder ( 1X PBS, pH 7.3,) | |
| Mouse | |
| >95% | |
| RUO | |
| Primary | |
| For reconstitution, we recommend adding 100uL distilled water to a final antibody concentration of about 1 mg/mL. To use this carrier-free antibody for conjugation experiment, we strongly recommend performing another round of desalting process. | |
| Store at -20 degrees C. Avoid freeze/thaw cycles. | |
| Lyophilized |
| Western Blot | |
| OTI2F2 | |
| Western Blot 1:2000 | |
| DNA-directed RNA polymerase II subunit F, DNA-directed RNA polymerases I, II, and III 14.4 kDa polypeptide, HRBP14.4, II, and III subunit RPABC2, polymerase (RNA) II (DNA directed) polypeptide F, RNA Polymerase II subunit 14.4 kD, RNA polymerases I, II, and III subunit ABC2, RPB6, RPB6 homolog | |
| Full length human recombiant protein of human POLR2F (NP_068809) produced in E.coli. | |
| 100 μg | |
| Core ESC Like Genes, Stem Cell Markers | |
| 5435 | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction