missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZCWPW2 Antibody (OTI10A12), Novus Biologicals Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-44790
This item is not returnable.
View return policy
Description
ZCWPW2 Monoclonal antibody specifically detects ZCWPW2 in Human samples. It is validated for Western Blot
Specifications
| ZCWPW2 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized powder ( 1X PBS, pH 7.3,) | |
| Mouse | |
| >95% | |
| RUO | |
| 152098 | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI10A12 | |
| Western Blot 1:2000 | |
| ZCW2, zinc finger CW-type PWWP domain protein 2, zinc finger, CW type with PWWP domain 2 | |
| Full length human recombinant protein of human ZCWPW2 (NP_001035522) produced in E.coli. | |
| 100 μg | |
| Primary | |
| For reconstitution, we recommend adding 100uL distilled water to a final antibody concentration of about 1 mg/mL. To use this carrier-free antibody for conjugation experiment, we strongly recommend performing another round of desalting process. | |
| Store at -20 degrees C. Avoid freeze/thaw cycles. | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction