missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD1d Antibody (1E3), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-42939
This item is not returnable.
View return policy
Description
CD1d Monoclonal antibody specifically detects CD1d in Human samples. It is validated for ELISA,Immunohistochemistry,Western Blot
Specifications
| CD1d | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.3, 50% glycerol | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA, Immunohistochemistry, Western Blot | |
| 1000 | |
| Western Blot 1:500 - 1:2000, ELISA, Immunohistochemistry 1:20 - 1:200 | |
| antigen-presenting glycoprotein CD1d, CD1A, CD1d antigen, CD1D antigen, d polypeptide, CD1d molecule, differentiation antigen CD1-alpha-3, HMC class I antigen-like glycoprotein CD1D, MGC34622, R3, R3G1, T-cell surface glycoprotein CD1d, thymocyte antigen CD1D | |
| CD1d molecule - (Uniprot# P15813) | |
| 100 μg | |
| Alzheimers Research, B Cell Development and Differentiation Markers, Cellular Markers, Mitotic Regulators, Myeloid derived Suppressor Cell, Neuroscience | |
| 912 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction