missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DBC1 Antibody (8C10), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-42956
This item is not returnable.
View return policy
Description
DBC1 Monoclonal antibody specifically detects DBC1 in Human,Mouse,Porcine samples. It is validated for ELISA,Western Blot,Immunocytochemistry/ Immunofluorescence
Specifications
| DBC1 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.3, 50% glycerol | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Mouse, Pig | |
| Purified |
| ELISA, Western Blot, Immunocytochemistry/Immunofluorescence | |
| 8C10 | |
| Western Blot 1:500 - 1:2000, ELISA, Immunocytochemistry/ Immunofluorescence 1:50 - 1:500 | |
| DBCCR1deleted in bladder cancer protein 1, deleted in bladder cancer 1, deleted in bladder cancer chromosome region candidate 1, FAM5AbA574M5.1 (deleted in bladder cancer chromosome region candidate 1 (IB3089A)), Protein FAM5A | |
| deleted in bladder cancer 1 - (Uniprot# O60477) | |
| 100 μg | |
| Breast Cancer, Cancer, Cell Cycle and Replication, p53 Pathway, Tumor Suppressors | |
| 1620 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction