missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EEF1D Antibody (6D9), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-42962
This item is not returnable.
View return policy
Description
EEF1D Monoclonal antibody specifically detects EEF1D in Human,Mouse samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunocytochemistry/ Immunofluorescence
Specifications
| EEF1D | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.3, 50% glycerol | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| ELISA, Immunohistochemistry, Western Blot, Immunocytochemistry/Immunofluorescence | |
| 6D9 | |
| Western Blot 1:500 - 1:2000, ELISA, Immunohistochemistry 1:50 - 1:500, Immunocytochemistry/ Immunofluorescence 1:10 - 1:100 | |
| Antigen NY-CO-4, EF-1D, EF-1-delta, EF1DFP1047, elongation factor 1-delta, eukaryotic translation elongation factor 1 delta (guanine nucleotide exchangeprotein), FLJ20897, guanine nucleotide exchange protein | |
| eukaryotic translation elongation factor 1 delta(guanine nucleotide exchange protein) - (Uniprot# P29692) | |
| 100 μg | |
| metabolism | |
| 1936 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction