missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PAX1 Antibody (8D9), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-43013
This item is not returnable.
View return policy
Description
PAX1 Monoclonal antibody specifically detects PAX1 in Human,Mouse samples. It is validated for ELISA,Western Blot
Specifications
| PAX1 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.3, 50% glycerol | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 5075 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG1 |
| ELISA, Western Blot | |
| 8D9 | |
| Western Blot 1:500 - 1:2000, ELISA | |
| paired box 1, paired domain gene HuP48 | |
| paired box 1 - (Uniprot# P15863) | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction