missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RABEP2 Antibody (2F7), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-43022
This item is not returnable.
View return policy
Description
RABEP2 Monoclonal antibody specifically detects RABEP2 in Human samples. It is validated for ELISA,Immunohistochemistry,Western Blot
Specifications
| RABEP2 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.3, 50% glycerol | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA, Immunohistochemistry, Western Blot | |
| 2F7 | |
| Western Blot 1:500 - 1:1000, ELISA, Immunohistochemistry 1:50 - 1:500 | |
| FLJ23282, FRA, rab GTPase-binding effector protein 2, rabaptin, RAB GTPase binding effector protein 2, Rabaptin-5 beta, rabaptin-5beta, RABPT5B | |
| rabaptin, RAB GTPase binding effector protein 2 - (Uniprot# Q9H5N1) | |
| 100 μg | |
| Cytokine Research | |
| 79874 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction