missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRUSS Antibody (1C0), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-43055
This item is not returnable.
View return policy
Description
TRUSS Monoclonal antibody specifically detects TRUSS in Human samples. It is validated for ELISA,Immunohistochemistry,Western Blot
Specifications
| TRUSS | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.3, 50% glycerol | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 26133 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG1 |
| ELISA, Immunohistochemistry, Western Blot | |
| 1C0 | |
| Western Blot 1:500 - 1:5000, ELISA, Immunohistochemistry 1:20 - 1:200 | |
| C20orf188, dJ756N5.2, DKFZp586C1223, DKFZP727M231, Protein TAP1, Protein TRUSS, short transient receptor potential channel 4 associated protein, short transient receptor potential channel 4-associated protein, TNF-receptor ubiquitous scaffolding/signaling protein, transient receptor potential cation channel, subfamily C, member 4 associatedprotein, Trp4-associated protein, Trpc4-associated protein, TRRP4APchromosome 20 open reading frame 188, TRUSS, tumor necrosis factor receptor-associated ubiquitous scaffolding and signalingprotein | |
| transient receptor potential cation channel, subfamily C, member 4 associated protein - (Uniprot# Q8TEL6) | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction