missing translation for 'onlineSavingsMsg'
Learn More

Annexin A6 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals NBP2-99419-100ul

 View more versions of this product

Product Code. 18645886

  • 326.00 € / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Annexin A6 Polyclonal antibody specifically detects Annexin A6 in Mouse samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Annexin A6
Polyclonal
Western Blot 1:500-1:2000
67 kDa calelectrin, annexin A6, Annexin VI, annexin VI (p68), annexin-6, ANX6, calcium-binding protein p68, calelectrin, calphobindin II, calphobindin-II, CBP68, Chromobindin-20, CPB-II, Lipocortin VI, p68, p70, Protein III
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human Annexin A6
100 μg
Endosome Markers
309
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG
Western Blot
Unconjugated
0.2 um filtered solution in PBS
Rabbit
Antigen and protein A Affinity-purified
RUO
Primary
Mouse
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.