missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
The FKBP10/FKBP65 antibody from Proteintech is a rabbit recombinant antibody generated with recombinant protein of human FKBP10/FKBP65. This antibody recognizes human antigen. The FKBP10/FKBP65 antibody has been validated for the following applications: WB, IHC, ELISA analysis.
Specifications
Specifications
| Antigen | FKBP10/FKBP65 |
| Applications | Western Blot, Immunohistochemistry, ELISA |
| Classification | Recombinant Monoclonal |
| Clone | S94 |
| Concentration | 500 μg/mL |
| Conjugate | Unconjugated |
| Gene Symbols | FKBP6; FKBP65; FLJ20683; FLJ22041; FLJ23833; hFKBP65 |
| Host Species | Rabbit |
| Immunogen | Recombinant Protein |
| Purification Method | Protein A purification |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?