missing translation for 'onlineSavingsMsg'
Learn More

DY131, MedChemExpress

Product Code. 16436568
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 16436568

Brand: Medchem Express HY15483

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress DY131 (GSK 9089) is a potent and selective ERRγ and ERRβ agonist. DY131displays inactive against ERRα, ERα and ERβ. DY131 also inhibits Smo signaling.

DY131 (GSK 9089) is a potent and selective ERRγ and ERRβ agonist. DY131displays inactive against ERRα, ERα and ERβ. DY131 also inhibits Smo signaling.

Specifications

Chemical Name or Material DY131
CAS 95167-41-2
Color Yellow
Purity Grade Notes Research
Molecular Formula C18H21N3O2
Quantity 10 mg
Synonym GSK 9089
Solubility Information DMSO : 50 mg/mL (160.58 mM; Need ultrasonic)
SMILES O=C(N/N=C/C1=CC=C(N(CC)CC)C=C1)C2=CC=C(O)C=C2
Molecular Weight (g/mol) 311.38
Formula Weight 311.38
Percent Purity 99.72%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.