missing translation for 'onlineSavingsMsg'
Learn More

OSI-930, MedChemExpress

Product Code. 16433698
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16433698 10 mg 10mg
16430087 50 mg 50mg
16426437 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16433698 Supplier Medchem Express Supplier No. HY10204

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress OSI-930 is an orally selective inhibitor of Kit, KDR and CSF-1R (c-Fms) with IC50s of 80 nM, 9 nM and 15 nM, respectively. OSI-930 also moderately inhibits Flt-1, c-Raf, Lck and low activity against PDGFRα/β, Flt-3 and Abl. OSI-930 has antitumor activity.

OSI-930 is an orally selective inhibitor of Kit, KDR and CSF-1R (c-Fms) with IC50s of 80 nM, 9 nM and 15 nM, respectively. OSI-930 also moderately inhibits Flt-1, c-Raf, Lck and low activity against PDGFRα/β, Flt-3 and Abl. OSI-930 has antitumor activity.

Specifications

Chemical Name or Material OSI-930
CAS 728033-96-3
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C22H16F3N3O2S
Quantity 10 mg
Solubility Information DMSO : 50 mg/mL (112.75 mM; Need ultrasonic)
SMILES O=C(C1=C(NCC2=CC=NC3=CC=CC=C23)C=CS1)NC4=CC=C(OC(F)(F)F)C=C4
Molecular Weight (g/mol) 443.44
Formula Weight 443.44
Percent Purity 98.13%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.