missing translation for 'onlineSavingsMsg'
Learn More

RO8994, MedChemExpress

Product Code. 16427258
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16427258

Brand: Medchem Express HY16999

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress RO8994 is a highly potent and selective series of spiroindolinone small-molecule MDM2 inhibitor, with IC50 of 5 nM (HTRF binding assays) and 20 nM (MTT proliferation assays).

RO8994 is a highly potent and selective series of spiroindolinone small-molecule MDM2 inhibitor, with IC50 of 5 nM (HTRF binding assays) and 20 nM (MTT proliferation assays).

Specifications

Chemical Name or Material RO8994
CAS 1309684-94-3
Color White
Purity Grade Notes Research
Molecular Formula C31H31Cl2FN4O4
Quantity 10 mg
Solubility Information DMSO : ≥ 45 mg/mL (73.35 mM)
SMILES O=C([C@H](N[C@H]1CC(C)(C)C)[C@H](C2=CC=CC(Cl)=C2F)[C@@]31C(NC4=C3C=CC(Cl)=C4)=O)NC5=CC=C(C(N)=O)C=C5OC
Molecular Weight (g/mol) 613.51
Formula Weight 613.51
Percent Purity 98.95%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.