missing translation for 'onlineSavingsMsg'
Learn More

TCN238, MedChemExpress

Product Code. 16476058
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16476058 10 mg 10mg
16496456 100 mg 100mg
16462197 50 mg 50mg
16438667 5 mg 5mg
16433426 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16476058 Supplier Medchem Express Supplier No. HY14419

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress TCN238 is an orally bioavailable mGlu4 receptor positive allosteric modulator (PAM) with an EC50 of 1 μM.

TCN238 is an orally bioavailable mGlu4 receptor positive allosteric modulator (PAM) with an EC50 of 1 μM.

Specifications

Chemical Name or Material TCN238
Physical Form Solid
CAS 125404-04-8
Color White
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C12H11N3
For Use With (Application) Neuroscience-Neuromodulation
Solubility Information DMSO : ≥ 150 mg/mL (760.49 mM)
SMILES NC1=NC=CC(/C=C/C2=CC=CC=C2)=N1
Molecular Weight (g/mol) 197.24
Formula Weight 197.24
Percent Purity 98.31%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.