missing translation for 'onlineSavingsMsg'
Learn More

TCS 359, MedChemExpress

Product Code. 16425908
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 16425908

Brand: Medchem Express HY13907

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress TCS 359, a 2-acylaminothiophene-3-carboxamide, is a potent and selective FLT3 inhibitor with an IC50 of 42 nM. TCS 359 inhibits MV4-11 cell proliferation with an IC50 of 340 nM.

TCS 359, a 2-acylaminothiophene-3-carboxamide, is a potent and selective FLT3 inhibitor with an IC50 of 42 nM. TCS 359 inhibits MV4-11 cell proliferation with an IC50 of 340 nM.

Specifications

Chemical Name or Material TCS 359
CAS 301305-73-7
Color White
Purity Grade Notes Research
Molecular Formula C18H20N2O4S
Quantity 10 mg
Solubility Information DMSO : 14.29 mg/mL (39.65 mM; Need ultrasonic)
SMILES O=C(C1=C(NC(C2=CC=C(OC)C(OC)=C2)=O)SC3=C1CCCC3)N
Molecular Weight (g/mol) 360.43
Formula Weight 360.43
Percent Purity 98.66%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.