missing translation for 'onlineSavingsMsg'
Learn More

11-epi-Prednisolone, TRC

Product Code. 30008023
Change view
Click to view available options
Quantity:
0.5 mg
5 mg
Unit Size:
0.5mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30008023 0.5 mg 0.5mg
30011101 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30008023 Supplier Toronto Research Chemicals Supplier No. TRCE5888550.5MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 600-90-8
Molecular Formula C21 H28 O5
Molecular Weight (g/mol) 360.44
Synonym (11α)-11,17,21-Trihydroxypregna-1,4-diene-3,20-dione, Pregna-1,4-diene-3,20-dione, 11α,17,21-trihydroxy- (8CI), 11α,17,21-Trihydroxy-1,4-pregnadiene-3,20-dione, Epiprednisolone, 11-epi-Prednisolone
IUPAC Name (8S,9S,10R,11R,13S,14S,17R)-11,17-dihydroxy-17-(2-hydroxyacetyl)-10,13-dimethyl-7,8,9,11,12,14,15,16-octahydro-6H-cyclopenta[a]phenanthren-3-one
SMILES C[C@]12C[C@@H](O)[C@H]3[C@@H](CCC4=CC(=O)C=C[C@]34C)[C@@H]1CC[C@]2(O)C(=O)CO

Specifications

Quantity 0.5 mg
InChI Formula InChI=1S/C21H28O5/c1-19-7-5-13(23)9-12(19)3-4-14-15-6-8-21(26,17(25)11-22)20(15,2)10-16(24)18(14)19/h5,7,9,14-16,18,22,24,26H,3-4,6,8,10-11H2,1-2H3/t14-,15-,16+,18+,19-,20-,21-/m0/s1
Formula Weight 360.1937
Percent Purity >95%
Chemical Name or Material 11-epi-Prednisolone
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.