missing translation for 'onlineSavingsMsg'
Learn More

12-Crown-4, 97%

Product Code. 10741414 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
2.5 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
2.5g
25g
5g
Product Code. Quantity Packaging unitSize
10741414 25 g Glass bottle 25g
10367840 2.5 g Glass bottle 2.5g
10317861 5 g Glass bottle 5g
3 options
This item is not returnable. View return policy

Product Code. 10741414

Brand: Thermo Scientific Acros 187020250

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 294-93-9 | C8H16O4 | 176.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chelation/Complexation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 294-93-9
Molecular Formula C8H16O4
Molecular Weight (g/mol) 176.21
MDL Number MFCD00005103
InChI Key XQQZRZQVBFHBHL-UHFFFAOYSA-N
PubChem CID 9269
ChEBI CHEBI:32399
IUPAC Name 1,4,7,10-tetraoxacyclododecane
SMILES C1COCCOCCOCCO1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 16°C
Color Colorless to Yellow
Density 1.0800g/mL
Boiling Point 238°C
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.4600 to 1.463
Quantity 25 g
Fieser 06,133; 09,126; 12,147; 13,167
Specific Gravity 1.08
Synonym 12-crown-4, eoct, 12-crown-4 ether, ethylene oxide cyclic tetramer, ccris 157, unii-k6069p2c2a, 12-crown4-ether, 12-crown 4-ether, 5-19-11-00334 beilstein handbook reference, 3,6,9,12-tetraoxacyclododecan
Solubility Information Solubility in water: miscible
Formula Weight 176.21
Percent Purity 97%
Physical Form Liquid After Melting
Chemical Name or Material 12-Crown-4
Show More Show Less
compliance-icons
Product Identifier
  • 12-Crown-4
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 1
Hazard Statement
  • H330-Fatal if inhaled.
Precautionary Statement
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.