accessibility menu, dialog, popup

UserName

OOOO

CAS RN 294-93-9

IUPAC Name: 1,4,7,10-tetraoxacyclododecane
Synonyms: EOCT 12-crown-4 ether ethylene oxide cyclic tetramer ccris 157
Molecular Weight (g/mol): 176.21
Molecular Formula: C8H16O4
InChi Key: XQQZRZQVBFHBHL-UHFFFAOYSA-N
SMILES: C1COCCOCCOCCO1
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
Boiling Point 
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)