missing translation for 'onlineSavingsMsg'
Learn More

12-Crown-4, 98%

Product Code. 11493994
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
Product Code. Quantity unitSize
11493994 25 g 25g
11473994 1 g 1g
11483994 5 g 5g
3 options
This item is not returnable. View return policy

Product Code. 11493994

Brand: Thermo Scientific Alfa Aesar A11972.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 294-93-9 | C8H16O4 | 176.21 g/mol

It serves as an aprotic solvent for alkali metals, the sodium solution is a strong reducing agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It serves as an aprotic ″solvent″ for alkali metals, the sodium solution is a strong reducing agent.

Solubility
Fully miscible in water.

Notes
Light & Air Sensitive & Hygroscopic. Store under inert gas. Keep in a tightly closed container.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 294-93-9
Molecular Formula C8H16O4
Molecular Weight (g/mol) 176.21
MDL Number MFCD00005103
InChI Key XQQZRZQVBFHBHL-UHFFFAOYSA-N
PubChem CID 9269
ChEBI CHEBI:32399
IUPAC Name 1,4,7,10-tetraoxacyclododecane
SMILES C1COCCOCCOCCO1

Specifications

Melting Point 17°C
Density 1.108
Boiling Point 68°C to 70°C (0.1 mmHg)
Flash Point >109°C (228°F)
Odor Odorless
Refractive Index 1.462
Quantity 25 g
UN Number UN2810
Beilstein 1363064
Sensitivity Air and light sensitive; Hygroscopic
Synonym 12-crown-4, eoct, 12-crown-4 ether, ethylene oxide cyclic tetramer, ccris 157, unii-k6069p2c2a, 12-crown4-ether, 12-crown 4-ether, 5-19-11-00334 beilstein handbook reference, 3,6,9,12-tetraoxacyclododecan
Solubility Information Fully miscible in water.
Formula Weight 176.22
Percent Purity 98%
Chemical Name or Material 12-Crown-4
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.