missing translation for 'onlineSavingsMsg'
Learn More

13-cis-N-[4-(Ethoxycarbonyl)phenyl]retinamide, MedChemExpress

Product Code. 16495406
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16495406 100 mg 100mg
16474558 10 mg 10mg
16427277 5 mg 5mg
16400867 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16495406 Supplier Medchem Express Supplier No. HY112077
 Restricted Item
Estimated Shipment: 01-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress 13-cis-N-[4-(Ethoxycarbonyl)phenyl]retinamide is a derivative of Retinoic acid.

13-cis-N-[4-(Ethoxycarbonyl)phenyl]retinamide is a derivative of Retinoic acid.

Specifications

Chemical Name or Material 13-cis-N-[4-(Ethoxycarbonyl)phenyl]retinamide
Physical Form Solid
CAS 251441-94-8
Color Light Yellow
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C29H37NO3
For Use With (Application) COVID-19-immunoregulation
Synonym Ethyl 4-((2Z,4E,6E,8E)-3,7-dimethyl-9-(2,6,6-trimethylcyclohex-1-en-1-yl)nona-2,4,6,8-tetraenamido)benzoate
Solubility Information DMSO : 10 mg/mL (22.34 mM; Need ultrasonic)
SMILES CC(/C=C/C=C(/C=C/C1=C(CCCC(C)1C)C)C)=C/C(NC2=CC=C(C(OCC)=O)C=C2)=O
Molecular Weight (g/mol) 447.61
Formula Weight 447.61
Percent Purity 98.11%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.