missing translation for 'onlineSavingsMsg'
Learn More
Learn More
14-3-3 Antibody (SR1457), Novus Biologicals™
Rabbit Monoclonal Antibody
349.00 € - 449.00 €
Specifications
| Antigen | 14-3-3 |
|---|---|
| Clone | SR1457 |
| Dilution | Western Blot 1:500-1:1000, Flow Cytometry 1:50, Immunohistochemistry 1:50-1:200 |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18604044
|
Novus Biologicals
NBP3-21694-100ul |
100 μg |
449.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18665474
|
Novus Biologicals
NBP3-21694-50ul |
50 μg |
349.00 €
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
14-3-3 Monoclonal antibody specifically detects 14-3-3 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, ImmunohistochemistrySpecifications
| 14-3-3 | |
| Western Blot 1:500-1:1000, Flow Cytometry 1:50, Immunohistochemistry 1:50-1:200 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| 14-3-3 protein tau, 14-3-3 protein T-cell, 14-3-3 protein theta, 14-3-3,1C5, HS1, Protein HS1, protein tau, tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein, thetapolypeptide | |
| A synthesized peptide derived from human 14-3-3 (Uniprot #: P31946, P63104) | |
| Primary | |
| Store at -20° C. Avoid freeze/thaw cycles. |
| SR1457 | |
| Western Blot, Flow Cytometry, Immunohistochemistry | |
| Unconjugated | |
| Rabbit | |
| Alzheimers Research, Cancer, Cell Biology, Cell Cycle and Replication, Growth and Development, Neuronal Cell Markers, Neuroscience, Phospho Specific, Protein Kinase, Signal Transduction | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| 10971 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title