missing translation for 'onlineSavingsMsg'
Learn More
Learn More
14-3-3 eta Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99490-100ul
This item is not returnable.
View return policy
Description
14-3-3 eta Polyclonal antibody specifically detects 14-3-3 eta in Human samples. It is validated for Western Blot, Immunoprecipitation
Specifications
| 14-3-3 eta | |
| Polyclonal | |
| Western Blot 1:500-1:2000, Immunoprecipitation 5-10 uL/mg of lysate | |
| 14-3-3 protein eta, Protein AS1, tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein, etapolypeptide, YWHA114-3-3 eta | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human 14-3-3 eta. | |
| 100 μg | |
| Apoptosis, Cancer, Neuroscience | |
| 7533 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunoprecipitation | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction