missing translation for 'onlineSavingsMsg'
Learn More
Learn More
14-3-3 zeta Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-24593
This item is not returnable.
View return policy
Description
14-3-3 zeta Polyclonal specifically detects 14-3-3 zeta in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin).
Specifications
| 14-3-3 zeta | |
| Polyclonal | |
| Unconjugated | |
| 0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide | |
| YWHAZ | |
| A portion of amino acids 200-245 of human 14-3-3 zeta was used as the immunogen for the antibody. | |
| 0.1 mg | |
| Cell Cycle and Replication | |
| 7534 | |
| Human, Mouse, Rat, Bovine, Canine, Primate | |
| Purified |
| Western Blot, Immunohistochemistry (Paraffin) | |
| 0.5 mg/mL | |
| Western Blot 1-3 ug/ml, Immunohistochemistry-Paraffin 10 ug/ml | |
| KCIP-1MGC126532,14-3-3-zeta, MGC111427, MGC138156, tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein, deltapolypeptide, tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein, zetapolypeptide, zeta polypeptide | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| The amino acid sequence used as immunogen for the antibody is 100% homologous in human and 94% homologous in mouse, rat and bovine. | |
| Store at -20C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction