Learn More
Corning™ 1536-Well Black/Clear Round Bottom Ultra-low Attachment Spheroid Microplate with Lid, Sterile
Product Code. 17770708 Shop All Corning Products
Change view
Click to view available options
Color:
Black, Clear Bottom
Unit Size:
Pack of 5
Product Code. Color unitSize
17770708 Black, Clear Bottom Pack of 5
1 options
Product Code. 17770708 Supplier Corning™ Supplier No. 4637
Only null left
Add to basket
Add to basket
With their novel and proprietary design, these microplates are ideal for generating and analyzing 3D multicellular spheroids in the same microplate. The Ultra-Low Attachment (ULA) surface enables uniform and reproducible 3D multicellular spheroid formation. The black opaque microplate body shields each optically clear, round bottom well from well-to-well cross-talk.
  • 1536-well format allows for high throughput 3D cell culture
  • Optically clear round bottom with black opaque microplate body
  • Covalent attachment of Ultra-Low Attachment surface to reduce cellular adhesion to well surface
  • Novel well geometry aids in the generation of uniform, single spheroids across all wells, which enables automated visualization.
  • Unique design shields each well to minimize well-to-well cross-talk.
  • You can culture and assay spheroids in the same microplate, without the need for transfer to a new microplate.
  • Compatible with the GNF ultra high throughput system
TRUSTED_SUSTAINABILITY
Specifications
Lid With Lid
Color Black, Clear Bottom
Volume (Metric) Well 14 mL
No. of Wells 1536
Volume (Metric) Working 5 to 10 μL
Sterility Sterile
Surface Treatment Ultra-low Binding
Well Shape Round Bottom (U-well)
Well Color Clear