Learn More
Corning™ 1536-Well Polystyrene NBS™-Treated Microplates
GREENER_CHOICE
Product Code. 10143613 Shop All Corning Products
Change view
Click to view available options
Color:
Black
Black, Clear Bottom
White
White, Clear Bottom
Includes:
Barcode
Generic barcodes
Without barcode
Unit Size:
Pack of 50
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Includes unitSize
10143613 Black Generic barcodes Pack of 50
17720708 White, Clear Bottom Barcode Pack of 50
10052443 White Generic barcodes Pack of 50
17790698 Black, Clear Bottom Barcode Pack of 50
10189472 Black, Clear Bottom Without barcode Pack of 50
10027322 Black Without barcode Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 10143613 Supplier Corning™ Supplier No. 3728BC
  • ONLINE EXCLUSIVE PRICE
    1585.00 €
    1479.54 € / Pack of 50
    Save 105.46 €
    6% Off
Estimated Shipment: 30-07-2026
Add to basket
Add to basket
Proprietary nonbinding surface is ideal for homogeneous assays with sticky enzymes or for reducing nonspecific binding to microplate wells
Corning 1536-well microplates are our highest density microplates available for high throughput screening. The microplates conform to standard microplate footprint and dimensions. These microplates are offered in solid black and white polystyrene, with round or flat bottoms, and in black-clear bottom formats.
  • Total well volume of 10 μL for round well microplates and 12.8 μL for flat bottom microplates
  • Recommended working volume up to 8 μL
  • Round well bottom for reduced air entrapment and improved CV values and Z factor
  • Raised well bottom for higher sensitivity
  • Flood reservoir on four sides to reduce instrument contamination
  • Lids are available separately. Corning lid (Cat. No. 3098) is compatible with these microplates.
Order Info
Packaged in bags of ten
TRUSTED_SUSTAINABILITY
Specifications
Format Standard
Lid Without Lid
Array 32 x 48
Packaging 5 inner packs of 10
Color Black
Product Type 1536-Well Cell Culture Multiwell Plates
Material Polystyrene
No. of Wells 1536
Volume (Metric) Working 8 μL
Sterility Non-sterile
Surface Treatment Non-binding
Total Volume (Metric) Well 12.5 μL
Well Shape Flat Bottom (F-well)
Includes Generic barcodes
No. per Case 50/Cs.
No. per Pack 10/Pk.
Quantity 50/Cs.
Well Color Black