Learn More
Corning™ 1536-Well White Polystyrene NBS Microplate without Lid, Non-sterile
Shop All Corning ProductsDescription
- Excellent signal dynamic range due to low background fluorescence and enhanced signal intensity
- Superior flatness minimizes variability within a plate and between plates
- Flat bottom wells are compatible with automated dispensers including the PerkinElmer FlexDrop™
- NBS microplates are ideal for homogeneous assays with enzymes or for reducing nonspecific binding to microplate wells
Specifications
Specifications
| Lid | Without Lid |
| Color | White |
| Material | Polystyrene |
| No. of Wells | 1536 |
| Volume (Metric) Working | Up to 8 μL |
| Sterility | Non-sterile |
| Surface Treatment | NBS |
| Quantity | 10/Pk., 50/Cs. |