missing translation for 'onlineSavingsMsg'
Learn More
Learn More
15b-Methyl Drospirenone, TRC
Specifications
Specifications
| Chemical Name or Material | 15beta-Methyl Drospirenone |
| Analyte or Component Names | 15beta-Methyl Drospirenone |
| Shipping Condition | Room Temperature |
| Molecular Formula | C24 H32 O3 |
| Synonym | 15beta-Methyl-3-oxo-6alpha,7alpha-dihydro-3′H-cyclopropa[6,7]-17alpha-pregn-4-ene-21,17-carbolactone, 15β-Methyl-3-oxo-6α,7α-dihydro-3′H-cyclopropa[6,7]-17α-pregn-4-ene-21,17-carbolactone, 3”3-16-Secodrospirenone, Drospirenone Imp. F (EP), 15β-Methyl Drospirenone, 15β-Methyl-6β,7β-methylene-3-oxo-17α-pregn-4-ene-21,17-carbolactone, 17-Hydroxy-15β-methyl-6β,7β-methylene-3-oxo-17α-pregn-4-ene-21-carboxylic Acid, Υ-Lactone, Drospirenone Impurity F |
| InChI Formula | InChI=1S/C24H32O3/c1-13-12-24(9-6-19(26)27-24)23(3)8-5-17-20(21(13)23)16-11-15(16)18-10-14(25)4-7-22(17,18)2/h10,13,15-17,20-21H,4-9,11-12H2,1-3H3/t13-,15-,16+,17+,20-,21+,22-,23+,24+/m1/s1 |
| Solution Type | Neat |
| SMILES | C[C@@H]1C[C@@]2(CCC(=O)O2)[C@@]3(C)CC[C@H]4[C@@H]([C@H]5C[C@H]5C6=CC(=O)CC[C@]46C)[C@H]13 |
| Industry Type | Pharmaceutical |
| Molecular Weight (g/mol) | 368.5091 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?