missing translation for 'onlineSavingsMsg'
Learn More

Tuttnauer 160L Benchtop Autoclave

Product Code. 17898988 Shop All Tuttnauer Products
Change view
Click to view available options
Features:
Three Phase
Three Phase, Cooling
Three Phase, Cooling, Vacuum, Steam Generator, BioHazard Filter (CL3 Waste), Printer
Three Phase, Cooling, Vacuum, Steam Generator, Printer
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Features unitSize
17898988 Three Phase, Cooling, Vacuum, Steam Generator, BioHazard Filter (CL3 Waste), Printer Each
17838998 Three Phase Each
17888988 Three Phase, Cooling Each
17858988 Three Phase, Cooling, Vacuum, Steam Generator, Printer Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 17898988 Supplier Tuttnauer Supplier No. 5075ELCPGBHP380D
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

The 5075EL-D has an advanced multi-color control panel and a chamber made of 316L stainless steel. Each model has a number of optional added-value features which can be configured for fast cooling, efficient drying, biohazard and waste sterilization, Fo control, and more.

Designed for your laboratory applications. The 5075EL-D has an advanced multi-color control panel and a chamber made of 316L stainless steel. Each model has a number of optional added-value features which can be configured for fast cooling, efficient drying, Fo control, and more.

  • Advanced Control System - Take Advantage of Tuttnauer’s state-of-the-art control system with multi-color display
  • Safe - Automatic pressure locking system. Door will not open when the chamber is overpressured
  • Optional Internal Printing - Documents date, time, temperature and pressure
  • Intuitive Multi-Color Display - Multi-color display for easier reading gives quick access to important information. Color is used to indicate the stage of the cycle
  • High Quality Stainless Steel - 316L grade stainless steel chamber and door with electro-polish finish
  • Diverse Load Types - The 5075EL-D autoclave comes with 8 factory set cycle programs that cover the processing needs of most laboratories and research centers

Specifications

Capacity (English) 35.2 gal.
Capacity (Metric) 160 L
Depth (Metric) Chamber 75 cm
Diameter (English) Chamber 20 in.
Depth (English) Chamber 30 in.
Diameter (Metric) Chamber 50 cm
Length (English) 47.2 cm
Length (Metric) 118 cm
Product Type Benchtop Autoclave
Temperature (English) 221°F to 279°F
Temperature (Metric) 105°C to 137°C
Features Three Phase, Cooling, Vacuum, Steam Generator, BioHazard Filter (CL3 Waste), Printer
Material 316L Stainless Steel
Sterility Non-sterile
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.