missing translation for 'onlineSavingsMsg'
Learn More

1A-116, MedChemExpress

Product Code. 16453948
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16453948 10 mg 10mg
16474966 100 mg 100mg
16442926 25 mg 25mg
16436667 5 mg 5mg
16410307 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16453948 Supplier Medchem Express Supplier No. HY104064

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress 1A-116, a potent Rac1 inhibitor, is specific for W56 residues, can prevent EGF-induced Rac1 activation and block Rac1-P-Rex1 interaction. 1A-116 can induce apoptosis and inhibit cell proliferation, migration and cycle progression in a concentration-dependent manner. 1A-116 also demonstrates a high antimetastatic activity in vivo.

1A-116, a potent Rac1 inhibitor, is specific for W56 residues, can prevent EGF-induced Rac1 activation and block Rac1-P-Rex1 interaction. 1A-116 can induce apoptosis and inhibit cell proliferation, migration and cycle progression in a concentration-dependent manner. 1A-116 also demonstrates a high antimetastatic activity in vivo.

Specifications

Chemical Name or Material 1A-116
Physical Form Solid
CAS 1430208-73-3
Color Off-White
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C16H16F3N3
For Use With (Application) Cancer-Kinase/protease
Solubility Information DMSO : 100 mg/mL (325.40 mM; Need ultrasonic)
SMILES N=C(NC1=CC=CC=C1C(F)(F)F)NC2=CC(C)=CC(C)=C2
Molecular Weight (g/mol) 307.31
Formula Weight 307.31
Percent Purity 99.39%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.