missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phthalic acid, bis-isoundecyl ester (mixture of branched chain isomers), Dr. Ehrenstorfer
Click to view available options
Quantity:
1 g
Unit Size:
1g
Specifications
Specifications
| Chemical Name or Material | Diisoundecyl Phthalate (Mixture of Branched Isomers) |
| CAS | 96507-86-7 |
| Molecular Formula | C30 H50 O4 |
| Quantity | 1 g |
| Synonym | 1,2-Benzenedicarboxylic Acid, Diisoundecyl Ester (9CI), DIUP, Jayflex DIUP, 1,2-Benzenedicarboxylic Acid, 1,2-Diisoundecyl Ester |
| InChI Formula | InChI=1S/C30H50O4/c1-25(2)19-13-9-5-7-11-17-23-33-29(31)27-21-15-16-22-28(27)30(32)34-24-18-12-8-6-10-14-20-26(3)4/h15-16,21-22,25-26H,5-14,17-20,23-24H2,1-4H3 |
| SMILES | CC(C)CCCCCCCCOC(=O)c1ccccc1C(=O)OCCCCCCCCC(C)C |
| IUPAC Name | bis(9-methyldecyl) benzene-1,2-dicarboxylate |
| Molecular Weight (g/mol) | 474.716 |
| Formula Weight | 474.371 g/mol |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction