missing translation for 'onlineSavingsMsg'
Learn More

1H-Benzotriazole, 99%

Product Code. 10145823 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Unit Size:
100g
2.5kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10145823 2.5 kg Plastic bottle 2.5kg
10429670 100 g Plastic bottle 100g
10235620 500 g Plastic bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10145823 Supplier Thermo Scientific Acros Supplier No. 183880025
 Restricted Item
Estimated Shipment: 03-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 95-14-7 | C6H5N3 | 119.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 95-14-7
Molecular Formula C6H5N3
Molecular Weight (g/mol) 119.13
MDL Number MFCD00005699
InChI Key QRUDEWIWKLJBPS-UHFFFAOYSA-N
Synonym 1h-benzotriazole, benzotriazole, 1,2,3-benzotriazole, 1h-benzo d 1,2,3 triazole, azimidobenzene, 1h-1,2,3-benzotriazole, aziminobenzene, benztriazole, benzene azimide, benzisotriazole
PubChem CID 7220
ChEBI CHEBI:75331
IUPAC Name 2H-benzotriazole
SMILES C1=CC2=NNN=C2C=C1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 94.0°C to 99.0°C
Color Beige-Yellow to White
Boiling Point 204.0°C (15.0 mmHg)
Flash Point 170°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Plastic bottle
Quantity 2.5 kg
Beilstein 26, 38
Fieser 16,21
Merck Index 15, 1111
Solubility Information Solubility in water: 25g/L in water (20°C). Other solubilities: soluble in alcohol,benzene,toluene,chloroform,and dmf,soluble in ethanol,methanol and ethyleneglycol
Formula Weight 119.13
Percent Purity 99%
Physical Form Powder, Granules, Crystals, Needles or Flakes
Chemical Name or Material 1H-Benzotriazole, 98%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1H-Benzotriazole
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H319-Causes serious eye irritation.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P273-Avoid release to the environment.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P391-Collect spillage.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: 1H-Benzotriazole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.