accessibility menu, dialog, popup

UserName

HNNN

CAS RN 95-14-7

IUPAC Name: 2H-1,2,3-benzotriazole
Synonyms: 2H-benzotriazole
Molecular Weight (g/mol): 119.13
Molecular Formula: C6H5N3
InChi Key: QRUDEWIWKLJBPS-UHFFFAOYSA-N
SMILES: N1N=C2C=CC=CC2=N1
Molecular Weight (g/mol) 
  • (2)
  • (4)
  • (3)
Percent Purity 
  • (1)
  • (2)
  • (6)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (1)
  • (2)
  • (3)
Physical Form 
  • (3)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (4)
  • (3)
  • (1)
  • (2)
  • (6)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (3)
  • (1)