missing translation for 'onlineSavingsMsg'
Learn More

1H-Indene, 97%

Product Code. 15498959
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
Product Code. Quantity unitSize
15498959 100 g 100g
11383296 5 g 5g
11393296 25 g 25g
3 options
This item is not returnable. View return policy

Product Code. 15498959

Brand: Thermo Scientific Alfa Aesar L12665.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 95-13-6 | C9H8 | 116.163 g/mol

1H-Indene is used in the synthesis of new C60 derivative, indene-C60 bisadduct1. It was used in preparing polyindene by the controlled cationic polymerization initiated with cumyl methyl ether/TiCl4 in CH2Cl2. It is also used in the synthesis of istatins and in the production of indene/cumarone thermoplastic resins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1H-Indene is used in the synthesis of new C60 derivative, indene-C60 bisadduct1. It was used in preparing polyindene by the controlled cationic polymerization initiated with cumyl methyl ether/TiCl4 in CH2Cl2. It is also used in the synthesis of istatins and in the production of indene/cumarone thermoplastic resins.

Solubility
Soluble in ethanol, acetone, ethyl ether, and ether. Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which areopened must be carefully resealed and kept upright to prevent leakage. Incompatible with strong oxidizing agents Recommended storage temperature is 2 - 8°C. It is sensitive to light.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 95-13-6
Molecular Formula C9H8
Molecular Weight (g/mol) 116.163
MDL Number MFCD00003777
InChI Key YBYIRNPNPLQARY-UHFFFAOYSA-N
Synonym indene, indonaphthene, inden, unii-67h8y6lb8a, indenyl radical, 1h-inden, pubchem20728, indene, reag, acmc-209ruy
PubChem CID 7219
ChEBI CHEBI:41921
IUPAC Name 1H-indene
SMILES C1C=CC2=CC=CC=C21

Specifications

Melting Point ∼-2°C
Density 0.996
Boiling Point 181°C to 182°C
Flash Point 58°C (136°F)
Odor Aromatic
Refractive Index 1.574
Quantity 100 g
UN Number UN3295
Beilstein 635873
Sensitivity Light sensitive
Merck Index 14,4939
Solubility Information Soluble in ethanol,acetone,ethyl ether,and ether. Insoluble in water.
Formula Weight 116.16
Percent Purity 97%
Chemical Name or Material 1H-Indene
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.