missing translation for 'onlineSavingsMsg'
Learn More

1H-Pyrazole-1-carboxamidine hydrochloride, 99%

Product Code. 15407118
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15407118 10 g 10g
15417118 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15407118 Supplier Thermo Scientific Alfa Aesar Supplier No. H60631.09
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 4023-02-3 | C4H6N4 | 110.12 g/mol

1H-Pyrazole-1-carboxamidine hydrochloride may be used in the following studies:

  • Preparation of guanidylated hollow fiber membranes.
  • SGuanylation of amines and in peptide synthesis.
  • Synthesis of bis-guanidinium-cholesterol derivatives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1H-Pyrazole-1-carboxamidine hydrochloride may be used in the following studies: • Preparation of guanidylated hollow fiber membranes. • Guanylation of amines and in peptide synthesis. • Synthesis of bis-guanidinium-cholesterol derivatives.

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents. Store under room temperature.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4023-02-3
MDL Number MFCD00210087

Specifications

Melting Point 167°C to 170°C
Molecular Formula C12H5ClO3
Quantity 10 g
Synonym 4-chloro-1,8-naphthalic anhydride, 4-chloronaphthalic anhydride, 6-chlorobenzo de isochromene-1,3-dione, 4-chloronapthalic anhydride, 6-chloro-1h,3h-benzo de isochromene-1,3-dione, 4-chloronaphthalene-1,8-dicarboxylic anhydride, 1h,3h-naphtho 1,8-cd pyran-1,3-dione, 6-chloro, naphthalic anhydride, 4-chloro-7ci,8ci, 6-chloro-1h,3h-naphtho 1,8-cd pyran-1,3-dione
Solubility Information Soluble in water.
InChI Key UJEUBSWHCGDJQU-UHFFFAOYSA-N
SMILES C1=CC2=C(C=CC3=C2C(=C1)C(=O)OC3=O)Cl
Molecular Weight (g/mol) 232.62
PubChem CID 77672
Formula Weight 146.58
Percent Purity 99%
Chemical Name or Material 1H-Pyrazole-1-carboxamidine hydrochloride
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 1H-Pyrazole-1-carboxamidine hydrochloride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 1
  • Skin sensitiser Category 1
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H373-May cause damage to organs.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : 1H-Pyrazole-1-carboxamidine hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.