missing translation for 'onlineSavingsMsg'
Learn More

ND-646, MedChemExpress

Product Code. 16430929
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16430929

Brand: Medchem Express HY101842

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress ND-646 is an orally bioavailable and steric inhibitor of acetyl-CoA carboxylase (ACC) with IC50s of 3.5 nM and 4.1 nM for recombinant hACC1 and hACC2, respectively.

ND-646 is an orally bioavailable and steric inhibitor of acetyl-CoA carboxylase (ACC) with IC50s of 3.5 nM and 4.1 nM for recombinant hACC1 and hACC2, respectively.

Specifications

Chemical Name or Material ND-646
CAS 1434639-57-2
Color White
Purity Grade Notes Research
Molecular Formula C28H32N4O7S
Quantity 1 mg
Solubility Information DMSO : ≥ 100 mg/mL (175.86 mM)
SMILES O=C(N)C(C)(C)N(C(N(C[C@@H](C1=CC=CC=C1OC)OC2CCOCC2)C3=C4C(C)=C(C5=NC=CO5)S3)=O)C4=O
Molecular Weight (g/mol) 568.64
Formula Weight 568.64
Percent Purity 98.03%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.