missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress Rituximab

Product Code. 16423079
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16423079 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16423079 Supplier MedChemExpress Supplier No. HYP9913

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Rituximab is an anti-CD20 chimeric monoclonal antibody used to treat certain autoimmune diseases and types of cancer.

Specifications

Antigen CD20 type 1
Quantity 1 mg
Target Species Human
Content And Storage Please store the product under the recommended conditions in the Certificate of Analysis.
Product Type Inhibitory Antibodies
Shipping Condition Shipping with dry ice
CAS 174722-31-7
Purity or Quality Grade 97.83%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.