missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(1R,2R)-2-PCCA hydrochloride, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
5mg
Description
(1R,2R)-2-PCCA hydrochloride is a diastereomer of 2-PCCA, and acts as a potent GPR88 receptor agonist, with an EC50 of 3 nM in cell-free assay, and 603 nM in cell assay.
Specifications
Specifications
| Chemical Name or Material | (1R,2R)-2-PCCA hydrochloride |
| Physical Form | Solid |
| CAS | 1609563-71-4 |
| Color | Deep Yellow |
| Quantity | 5 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C30H39Cl2N3O |
| For Use With (Application) | Neuroscience-Neuromodulation |
| Solubility Information | DMSO : 50 mg/mL (94.60 mM; Need ultrasonic) |
| SMILES | CCCC1=CC=C(C2=CC=C(N(C[C@@H](N)[C@@H](C)CC)C([C@H]3[C@H](C4=CC=CC=N4)C3)=O)C=C2)C=C1.[H]Cl.[H]Cl |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction