missing translation for 'onlineSavingsMsg'
Learn More

(1R,2R)-N-(p-Toluenesulfonyl)-1,2-diphenylethanediamine, 98+%

Product Code. 11351999
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11351999 25 g 25g
11331999 1 g 1g
11341999 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11351999 Supplier Thermo Scientific Alfa Aesar Supplier No. H26061.14
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 144222-34-4 | C21H23N2O2S | 367.49 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 144222-34-4
Molecular Formula C21H23N2O2S
Molecular Weight (g/mol) 367.49
MDL Number MFCD02093428
InChI Key UOPFIWYXBIHPIP-NHCUHLMSSA-O
Synonym 1r,2r---n-4-toluenesulfonyl-1,2-diphenylethylenediamine, 1r,2r---n-p-tosyl-1,2-diphenylethylenediamine, r,r-tsdpen, r,r-n-p-toluenesulfonyl-1,2-diphenylethylenediamine, 1r,2r-n-p-tosyl-1,2-diphenylethylenediamine, n-1r,2r-2-amino-1,2-diphenylethyl-4-methylbenzenesulfonamide, r,r-n-2-amino-1,2-diphenylethyl-p-toluenesulfonamide, 1r,2r---n-p-tosyl-1,2-diphenylethylene diamine, 1r, 2r---n-4-toluene sulfonyl 1,2-diphenyl-1,2-ethane diamine, benzenesulfonamide, n-1r,2r-2-amino-1,2-diphenylethyl-4-methyl
PubChem CID 2734565
IUPAC Name N-[(1R,2R)-2-amino-1,2-diphenylethyl]-4-methylbenzenesulfonamide
SMILES CC1=CC=C(C=C1)S(=O)(=O)N[C@@H]([C@H]([NH3+])C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 127°C to 132°C
Quantity 25 g
Optical Rotation −31° (c=1 in Chloroform)
Formula Weight 366.48
Percent Purity ≥98%
Chemical Name or Material (1R,2R)-N-(p-Toluenesulfonyl)-1,2-diphenylethanediamine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.