missing translation for 'onlineSavingsMsg'
Learn More

2-(1-Naphthyl)-5-phenyloxazole, laser grade and suitable for scintillation spectrometry, 99+%

Product Code. 11409230
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11409230 1 g 1g
11419230 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11409230 Supplier Thermo Scientific Alfa Aesar Supplier No. A16679.03
 Restricted Item
Estimated Shipment: 08-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 846-63-9 | C19H13NO | 271.319 g/mol

Starting with the organic fluor 2-(1-naphthyl)-5-phenyloxazole (αNPO), the vNPO and allylNPO monomers were synthesized using Stille coupling followed by purification. Used as a laser dye.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Starting with the organic fluor 2-(1-naphthyl)-5-phenyloxazole (αNPO), the vNPO and allylNPO monomers were synthesized using Stille coupling followed by purification. Used as a laser dye.

Solubility
Sparingly Soluble (4.1E-4 g/L) (25°C). Soluble most organic solvents.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 846-63-9
Molecular Formula C19H13NO
Molecular Weight (g/mol) 271.319
MDL Number MFCD00019714
InChI Key WWVFJJKBBZXWFV-UHFFFAOYSA-N
Synonym 2-1-naphthyl-5-phenyloxazole, alpha-npo, oxazole, 2-1-naphthalenyl-5-phenyl, 2-1-naphthyl-5-phenyl-1,3-oxazole, 2-naphthalen-1-yl-5-phenyloxazole, oxazole, 2-1-naphthyl-5-phenyl, .alpha.-npo, anpo, 2-alpha-naphthyl-5-phenyloxazole, 2-naphthalen-1-yl-5-phenyl-1,3-oxazole
PubChem CID 70058
IUPAC Name 2-naphthalen-1-yl-5-phenyl-1,3-oxazole
SMILES C1=CC=C(C=C1)C2=CN=C(O2)C3=CC=CC4=CC=CC=C43

Specifications

Melting Point 104°C to 105°C
Quantity 1 g
Beilstein 210666
Solubility Information Sparingly Soluble (4.1E-4g/L) (25°C). Soluble most organic solvents.
Formula Weight 271.32
Percent Purity ≥99%
Grade Suitable for scintillation spectrometry
Chemical Name or Material 2-(1-Naphthyl)-5-phenyloxazole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.