missing translation for 'onlineSavingsMsg'
Learn More

2-(1-Naphthylmethyl)-2-imidazoline hydrochloride, 99%

Product Code. 11471177
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11471177 500 g 500g
11451177 25 g 25g
11461177 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11471177 Supplier Thermo Scientific Alfa Aesar Supplier No. B24161.36
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 550-99-2 | C14H15ClN2 | 246.738 g/mol

Naphazoline hydrochloride produces ionotropic effects via α-adrenoceptor stimulation. It is an activator of AR and Nischarin. It is used as a nasal decongestant and ocular vasoconstrictor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in water 170 g/L, alcohol, DMSO, ethanol.

Notes
Store in a cool, dry place in tightly closed container. Store at room temperature. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 550-99-2
Molecular Formula C14H15ClN2
Molecular Weight (g/mol) 246.738
MDL Number MFCD00012554
InChI Key DJDFFEBSKJCGHC-UHFFFAOYSA-N
Synonym naphazoline hydrochloride, albalon, naphazoline hcl, rhinantin, rhinoperd, stricylon, naphcon, niazol, rinofug, vasocon
PubChem CID 11079
ChEBI CHEBI:7470
IUPAC Name 2-(naphthalen-1-ylmethyl)-4,5-dihydro-1H-imidazole;hydrochloride
SMILES C1CN=C(N1)CC2=CC=CC3=CC=CC=C32.Cl

Specifications

Melting Point 257°C to 261°C
Quantity 500 g
UN Number UN2811
Beilstein 3716843
Merck Index 14,6368
Solubility Information Soluble in water 170g/L,alcohol,DMSO,ethanol.
Formula Weight 246.74
Percent Purity 99%
Chemical Name or Material 2-(1-Naphthylmethyl)-2-imidazoline hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.