accessibility menu, dialog, popup

UserName

HClNNH

CAS RN 550-99-2

IUPAC Name: hydrogen 2-[(naphthalen-1-yl)methyl]-4,5-dihydro-1H-imidazole chloride
Synonyms: naphazoline hydrochloride naphazoline (inn) and its salts
Molecular Weight (g/mol): 246.74
Molecular Formula: C14H15ClN2
InChi Key: DJDFFEBSKJCGHC-UHFFFAOYSA-N
SMILES: [H+].[Cl-].C(C1=NCCN1)C1=C2C=CC=CC2=CC=C1
Molecular Weight (g/mol) 
  • (3)
  • (8)
Quantity 
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
Percent Purity 
  • (3)
  • (5)
Physical Form 
  • (2)
  • (3)
Color 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results