missing translation for 'onlineSavingsMsg'
Learn More

2-(1-Piperazinyl)phenol, 98%

Product Code. 11409736
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11409736 10 g 10g
11419736 50 g 50g
11429736 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11409736 Supplier Thermo Scientific Alfa Aesar Supplier No. B20252.09
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 1011-17-2 | C10H14N2O | 178.235 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1011-17-2
Molecular Formula C10H14N2O
Molecular Weight (g/mol) 178.235
MDL Number MFCD00190246
InChI Key UORNTHBBLYBAJJ-UHFFFAOYSA-N
Synonym 2-piperazin-1-yl phenol, 1-2-hydroxyphenyl piperazine, 2-1-piperazinyl phenol, 2-1-piperazino phenol, o-1-piperazinyl phenol, 2-piperazin-1-yl-phenol, n-2-hydroxyphenyl piperazine, phenol, 2-1-piperazinyl, 1-2-hydroxyphenyl-piperazine, 2-piperazinylphenol
PubChem CID 70530
IUPAC Name 2-piperazin-1-ylphenol
SMILES C1CN(CCN1)C2=CC=CC=C2O

Specifications

Melting Point 122°C to 127°C
Quantity 10 g
UN Number UN3263
Beilstein 10646
Sensitivity Air Sensitive
Formula Weight 178.24
Percent Purity 98%
Chemical Name or Material 2-(1-Piperazinyl)phenol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.