missing translation for 'onlineSavingsMsg'
Learn More

2-(2,2,2-Trimethylacetamido)pyridine-3-boronic acid pinacol ester, 98%, Thermo Scientific Chemicals

Product Code. 11434380
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11434380 250 mg 250mg
11444380 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11434380 Supplier Thermo Scientific Alfa Aesar Supplier No. H50015.MD
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

2-(2,2,2-Trimethylacetamido)pyridine-3-boronic acid pinacol ester is used as an intermediate in organic synthesis. Further, it is used in agrochemicals, dyestuffs and in pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-(2,2,2-Trimethylacetamido)pyridine-3-boronic acid pinacol ester is used as an intermediate in organic synthesis. Further, it is used in agrochemicals, dyestuffs and in pharmaceuticals.

Notes
Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 532391-30-3
Molecular Formula C16H25BN2O3
Molecular Weight (g/mol) 304.20
MDL Number MFCD06658997
InChI Key JTGWGXPOGSUYCT-UHFFFAOYSA-N
Synonym n-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridin-2-yl pivalamide, 2,2-dimethyl-n-3-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-pyridin-2-yl-propionamide, 2-2,2,2-trimethylacetamido pyridine-3-boronic acid pinacol ester, 2,2-dimethyl-n-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridin-2-yl propanamide, 2-pivalamidopyridin-3-ylboronic acid pinacol ester, 2-2,2,2trimethylacetamido pyridine-3-boronic acid pinacol ester, 2,2-dimethyl-n-3-tetramethyl-1,3,2-dioxaborolan-2-yl pyridin-2-yl propanamide, 2,2-dimethyl-n-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-pyridin-2-yl-propionamide, acmc-20aojk
PubChem CID 17750164
IUPAC Name 2,2-dimethyl-N-[3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-yl]propanamide
SMILES CC(C)(C)C(=O)NC1=NC=CC=C1B1OC(C)(C)C(C)(C)O1

Specifications

Melting Point 155°C
Quantity 250 mg
Formula Weight 304.2
Percent Purity 98%
Chemical Name or Material 2-(2,2,2-Trimethylacetamido)pyridine-3-boronic acid pinacol ester

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.