missing translation for 'onlineSavingsMsg'
Learn More

2,2,2,3'-Tetrafluoroacetophenone, 98%

Product Code. 15402019
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15402019 1 g 1g
15412019 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15402019 Supplier Thermo Scientific Alfa Aesar Supplier No. H64704.03
 Restricted Item
Estimated Shipment: 12-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 708-64-5 | C8H4F4O | 192.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 708-64-5
Molecular Formula C8H4F4O
Molecular Weight (g/mol) 192.11
MDL Number MFCD00039236
InChI Key NQSPTMFCJGKOQJ-UHFFFAOYSA-N
Synonym 2,2,2,3'-tetrafluoroacetophenone, 2,2,2-trifluoro-1-3-fluorophenyl ethanone, m-fluorotrifluoroacetophenone, acetophenone, 2,2,2,3'-tetrafluoro, 2,2,2-trifluoro-1-3-fluorophenyl ethan-1-one, ethanone, 2,2,2-trifluoro-1-3-fluorophenyl, ethanone,2,2,2-trifluoro-1-3-fluorophenyl, acmc-20aol4, m,alpha,alpha,alpha-tetrafluoroacetophenone, 2,2,2,3'-trtrafluoroacetophenone
PubChem CID 69723
IUPAC Name 2,2,2-trifluoro-1-(3-fluorophenyl)ethanone
SMILES FC1=CC=CC(=C1)C(=O)C(F)(F)F

Specifications

Color Colorless to Yellow
Density 1.372 g/mL
Boiling Point 79°C (50 mmHg)
Flash Point 17°C (63°F)
Refractive Index 1.444
Quantity 1 g
UN Number UN1224
Beilstein 1872788
Formula Weight 192.11
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2,2,2,3'-Tetrafluoroacetophenone
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.